Would you like to log in again?
Serological Pipettes, Sarstedt
Supplier: Sarstedt
86.1254.001 86.1253.001 86.1251.001 86.1254.025 86.1253.025 86.1685.001 86.1251.025 86.1252.025 86.1252.001
101093-962CS 324.31 USD InStock
101093-962 101093-952 103575-136 101093-976 101093-966 103575-140 103575-138 101093-960 101093-972
Serological Pipettes, Sarstedt
Pipettes Serological Pipettes
These are sterile, plugged, graduated pipettes and are compatible with all common pipettors.
  • Easy identification by international code
  • Easy opening and static free packaging
The serological pipettes are manufactured from clear polystyrene and can be used for all suitable kind of applications. Individually wrapped pipettes feature larger pipetting volume due to negative graduation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty