Would you like to log in again?
Venous Blood Collection Tubes, Paper Label, Sarstedt
Supplier: Sarstedt
41.1393.105 41.1395.105
101094-002PK 38.32 USD InStock
101094-002 101094-004
Venous Blood Collection Tubes, Paper Label, Sarstedt
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Blood collection tubes which use the gravity-flow principle of collection are ideal in combination with the micro needles for blood collection in infants or newborns
The closure contains integral O-ring and skirted conical base. They are graduated and also have paper label with shelf life. The tubes are manufactured from clear polypropylene and are available with screw cap
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty