Would you like to log in again?
Surgi-OR™ Rochester-Pean Forceps, Mid-Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-498 95-487
10184-616EA 59.63 USD InStock
10184-616 10184-614
Surgi-OR™ Rochester-Pean Forceps, Mid-Grade, Sklar
Forceps
Surgi-OR™ Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty