Would you like to log in again?
Lysol® Quaternary Disinfectant Cleaner
Supplier: T. Frank McCall's
44103LY74983
21899-598CS 648.71 USD InStock
21899-598
Lysol® Quaternary Disinfectant Cleaner
Cleaning Agents Disinfectants
Quaternary disinfectant cleaner that has virucidal, fungicidal and bactericidal properties.
  • Effective for hospital disinfectant cleans that disinfects hard and non-porous surfaces
  • Economical and concentrated formula for effective cleaning
  • Multi-purpose, neutral pH, germicidal detergent and deodorant
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty