Would you like to log in again?
Vernier® Standard Cart
Supplier: Vernier
DTS-CART-S
470219-344EA 72.99 USD InStock
470219-344
Vernier® Standard Cart
Carts
This low friction cart is designed to study motion and collisions on the Vernier Combination 1.2 m Track/Optics Bench.
The Standard Cart has been redesigned to include:
Four mass trays to easily vary the total cart mass.
Anti-roll peg to ensure that the cart rolls on the track but not off the table.
Top tray for easy attachment of sensors
Vernier carts are also compatible with other common carts and tracks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty