Would you like to log in again?
Solar Energy Exploration Kit
Supplier: VERNIER SOFTWARE & TECH SE
KW-SEEK
470219-360EA 88.99 USD InStock
470219-360
Solar Energy Exploration Kit
Power Supplies
Explore solar energy with this innovative science kit designed to help students investigate energy transformations. A hinged box helps students discover how the angle of photovoltaic panels relative to the sun affects power output. Compare your experimental findings to online sun-angle calculators.
Three solar panels allow you to explore series and parallel circuits. Learn about the effect of important variables in photovoltaic systems, such as the effects of shade, temperature, and load on solar panel output. Harness energy from the solar panels to pump water or power a small motor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty