Would you like to log in again?
pH Buffer Capsules, 3×10
Supplier: VERNIER SOFTWARE & TECH SE
PH-BUFCAP
470219-362EA 33.99 USD InStock
470219-362
pH Buffer Capsules, 3×10
Buffers pH Calibration Buffers
The pH Buffer Capsules (3x10) kit is a convenient set of three vials containing capsules to make pH 4, pH 7, and pH 10 buffer standards for calibrating pH sensors. Each vial contains 10 capsules, each of which produces 100 mL of solution when mixed with DI water. Add three drops of the buffer preservative to 100 mL of your buffer solution to prolong the shelf life. Hydrion Color Key Buffer Preservative is a combination mold inhibitor and universal indicator for color coding the buffer solutions: orange for pH 4, green for pH 7, and blue for pH 10.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty