Would you like to log in again?
Lysyl Endopeptidase
Supplier: Fujifilm Wako Chemicals USA
125-05061
101974-094EA 1078.85 USD InStock
101974-094
Lysyl Endopeptidase
Enzymes
Freeze dried product that retains sufficient activity for in-gel digestion and packed in very small quantities .
  • High specificity and efficiency of protein digestion allow for easy database searches by peptide mass
  • Improved cleavage at lysine residue and increase in number of peptides obtained in combination with trypsin
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty