Would you like to log in again?
Demonstration Electrophorus
Supplier: EISCO SCIENTIFIC LLC
PH0871B
470219-520EA 36.85 USD InStock
470219-520
Demonstration Electrophorus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Comprised of a thick plastic base with four rubber feet, along with with an 18cm diameter aluminium plate with an insulating handle.
Produce a strong spark repeatedly, without recharging. The base is thick plastic with four rubber feet attached for convenient handling. The plastic material produces a strong frictional charge when rubbed with fur or other material. The 18cm diameter aluminum plate is designed to eliminate any sharp edges which could cause leakage. An insulating handle is attached.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty