Would you like to log in again?
Van de Graaff, 220 V
Supplier: EISCO SCIENTIFIC LLC
PH0920AV2
470219-528EA 374.65 USD InStock
470219-528
Van de Graaff, 220 V
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Specially designed for electrostatic experiments and where continuous source of high voltage is required. Fitted on base with smooth running DC Motor, operates on 220 volts A.C. 50 Hz. Charge collecting Belt: Silicone rubber having excellent insulation resistance. Tracking: Belt-tracking easily adjusted. Charge collecting combs: Aluminium mesh, clearly visible. Spheres: Aluminium 150mm. dia. Connections: 4mm sockets in dome and base. Voltage developed: Upto 200 kV, depending on ambient conditions and humidity, with a spark length of 60mm. Discharger: Spherical 100mm. dia., with insulated handle and 4mm sockets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty