Would you like to log in again?
High Traffic Series Medium Roll, Brady
Supplier: Brady
107718
10809-784RL 438.6 USD InStock
10809-784
High Traffic Series Medium Roll, Brady
Spill Control Materials
Built for durability and high absorbency, the economical "High Traffic" or HT Roll is ideal for your busy problem areas
The HT Roll will quickly become your mat of choice for saving time and money on your industrial application. 2-ply, gray, dimpled, medium weight high traffic roll. Perfed every 18" and up the center. Polypropylene material. Gray color.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty