Would you like to log in again?
MAXX Chemical Roll, Brady®
Supplier: Brady
134360
10796-290PK 456.42 USD InStock
10796-290
MAXX Chemical Roll, Brady®
Spill Control Materials
MAXX means greater performance without the extra weight
Specially developed chemical sorbent, yellow color-coded for safety and easy separation of hazardous waste, medium-weight pad. Does not react with aggressive fluids. Perfed every 15" and up the center.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty