Would you like to log in again?
Sklar Blue™ Pederson Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
91-5042 91-5046 91-5038 91-5047 91-5050
10184-386EA 488.41 USD InStock
10184-386 10184-392 10184-382 10194-112 10184-390
Sklar Blue™ Pederson Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Blue™ Pederson Vaginal Speculum is used for electrocsurgical gynecological procedures, such as LEEP.
It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The blades of the Pederson Speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient.
Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue.
Sklar Blue™ instruments are fully autoclavable and may also be cleaned ultrasonically. All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.
Delivery information: The Sklar Blue™ Graves Vaginal Speculum is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty