Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sklar Blue™ Mini Emmett Hook, OR Grade, Sklar
Supplier: Sklar
91-5246
10184-498EA 399.38 USD InStock
10184-498
Sklar Blue™ Mini Emmett Hook, OR Grade, Sklar
Retractors
This is also be known as an Emmett iris hook, is most commonly used in electrosurgical OB/GYN procedures involving the uterus.
  • The hook is at the end of a long, narrow handle
  • Feature high quality stainless steel with an insulated coating
This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue™ instrument is fully autoclavable and may also be cleaned ultrasonically.
Sklar Blue™ coated instrument comply with biological testing requirements for cytotoxicity and sensitization regulatory. This also comply with the pre-vacuum and gravity cycle steam sterilization validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coating are guaranteed for one year against manufacturing defects.
Certifications: Meets ISO 10993 requirements.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty