Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BLMH Mouse Monoclonal Antibody [clone: 760239]

Supplier: R & D Systems
MAB6200
102886-288EA 605.94 USD InStock
102886-288
Anti-BLMH Mouse Monoclonal Antibody [clone: 760239]
Antibodies
Type: Primary
Antigen: BLMH
Clonality: Monoclonal
Clone: 760239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty