Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIN2B Rabbit Polyclonal Antibody

Supplier: R & D Systems
PPS013
220022-125EA 663.78 USD InStock
220022-125
Anti-GRIN2B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: GRIN2B
Antigen Name: glutamate receptor, ionotropic, N-methyl D-aspartate 2B
Antigen Synonyms: EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty