Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Blue™ Bozeman Forceps, OR Grade, Sklar
Supplier: Sklar
91-5310 91-5308 91-5307
10184-522EA 374.55 USD InStock
10184-522 10184-524 10184-526
Blue™ Bozeman Forceps, OR Grade, Sklar
Forceps
Sklar Blue™ Bozeman Forceps are a multi-functional instrument commonly used in gynecological procedures
They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity. The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. The jaws are available either curved or straight. Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue™ instruments are fully autoclavable and may also be cleaned ultrasonically. All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty