Would you like to log in again?
Sklar Black™ Freeway-Graves Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3815 90-3755 90-3754 90-3814
10197-212EA 555.45 USD InStock
10197-212 10197-210 10197-228 10197-226
Sklar Black™ Freeway-Graves Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Black™ freeway-graves vaginal speculum is used for laser gynecological procedures.
It is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation. The Freeway-Graves Vaginal Speculum features an open side which allows improved visibility and more room for supplemental instrument access.
It is available with 45 or 90 degree angled blades. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty