Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
StemBoost™ Nociceptor Induction Cocktail Set (1000X), Sterile-Filtered, Biovision
Supplier: BioVision
AB286233-500UL AB286233-1
10013-090EA 283.43 USD InStock
10013-090 10013-088
StemBoost™ Nociceptor Induction Cocktail Set (1000X), Sterile-Filtered, Biovision
Cell Culture Supplements and Additives
A convenient, sterile-filtered cocktail solution (1000X, in DMSO) containing three small molecules useful to induce rapid and efficient neuronal differentiation of human pluripotent stem cells (hPSCs) into nociceptors. The three products are: GSK-3beta inhibitor CHIR99021 (Cat. No. 1677), gamma-secretase inhibitor DAPT (Cat. No. 1855) and FGFR inhibitor SU-5402 (Cat. No. 1645).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty