Would you like to log in again?
Lister Bandage Scissors, OR Grade, Sklar
Supplier: Sklar
22-1054 22-1072
89025-210EA 176.17 USD InStock
89025-210 89025-200
Lister Bandage Scissors, OR Grade, Sklar
Scissors
Sklar's® Lister Bandage Scissors are a versatile instrument with multiple uses
The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger.
They are commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric and occasionally umbilical cord.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty