Would you like to log in again?
Surgi-OR™ Crile Hemostatic Forceps, Mid Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-452 95-447 95-458 95-463
89023-626EA 46.93 USD InStock
89023-626 89023-628 89023-622 89023-624
Surgi-OR™ Crile Hemostatic Forceps, Mid Grade, Sklar
Forceps
Surgi-OR™ Crile Hemostatic Forceps, also known as a Crile Hemostat, are used for clamping blood vessels or tissue before cauterization or ligation.
Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique. They may also be used for soft tissue dissection typically no deeper than 6-8" or in Laparotomy procedures.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty