Would you like to log in again?
Mouse Recombinant FGF21 (from E. coli)
Supplier: BioVision
AB285477-50UG AB285477-10UG AB285477-1MG
10005-790EA 15137.8 USD InStock
10005-790 10005-788 10005-792
Mouse Recombinant FGF21 (from E. coli)
Proteins and Peptides
A heparin binding growth factor, stimulating the proliferation and activation of cells expressing FGFR
The FGFs are a family of more than 20 small (~17–26 kDa) secreted peptides. The initial characterization of these proteins focused on their ability to stimulate fibroblast proliferation. FGFs modulate cellular activity via at least 5 distinct subfamilies of high-affinity FGF receptors (FGFRs): FGFR-1, -2, -3, and -4, all with intrinsic tyrosine kinase activity and, except for FGFR-4, multiple splice isoforms, and FGFR-5, which lacks an intracellular kinase domain. There is growing evidence that FGFRs can be important for regulation of glucose and lipid homeostasis. FGFR-2 appears to be a key molecule during pancreatic development and FGFR-4 has been implicated in cholesterol metabolism and bile acid synthesis. FGF-21 is preferentially expressed in liver, but an exact knowledge of FGF-21 bioactivity and its mode of action have been lacking to date. FGF-21 is a potent activator of glucose uptake on adipocytes, protects animals from diet-induced obesity when overexpressed in transgenic mice, and lowers blood glucose and triglyceride levels when therapeutically administered to diabetic rodents. Fibroblast Growth Factor-21 Mouse Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 183 amino acids including N-terminal Methionine and having a molecular mass of 20.1 kDa. The amino acid sequence of the recombinant mouse FGF21 is 100% homologous to the amino acid sequence of the mouse FGF21 without signal sequence.
The FGF-21 is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty