Would you like to log in again?
Mouse Recombinant FLT3 Ligand (from E. coli)
Supplier: BioVision
AB51985-10
10005-804EA 236.07 USD InStock
10005-804
Mouse Recombinant FLT3 Ligand (from E. coli)
Proteins and Peptides
A member of a small family of growth factors that stimulate the proliferation of hematopoietic cells
Determined by the dose dependent proliferation of mouse AML5 cells
Flt3-Ligand belongs to a small group of growth factors that regulate proliferation of early hematopoietic cells. Multiple isoforms of Flt3-Ligand have been identified. The predominant form is the transmembrane which is biologically active on the cell surface. The proteolytically cleaved transmembrane isoform generates a soluble form, which is also biologically active. Flt3-Ligand binds to cells expressing the tyrosine kinase receptor Flt3. Flt3-Ligand alone cannot stimulate proliferation, but synergizes well with other CSFs and interleukins to induce growth and differentiation. Recombinant murine flt3-Ligand is a 16.4 kDa protein containing 144 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty