Would you like to log in again?
Human Recombinant GMCSF (from E. coli)
Supplier: BioVision
AB285486-1MG AB285486-10UG AB285486-100UG
10005-832EA 1596.31 USD InStock
10005-832 10005-834 10005-830
Human Recombinant GMCSF (from E. coli)
Proteins and Peptides
A cytokine that stimulates the bone marrow to produce granulocytes and stem cells
The ED₅₀ determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is <0.1 ng/ml, corresponding to a specific activity of >1 x 10⁷ units/mg.
Human GM-CSF (Granulocyte Macrophage Colony stimulating Factor) is a potent species-specific stimulator of bone marrow cells. GM-CSF stimulates precursor cells of granulocytes, macrophages and eosinophils. The human and murine molecules are species-specific and exhibit no cross-species reactivity. Recombinant human GM-CSF is a 14.6 kDa globular protein containing 128 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty