Would you like to log in again?
Surgi-OR™ Babcock Forceps, Mid Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-952 95-956
10184-632EA 79.1 USD InStock
10184-632 89023-744
Surgi-OR™ Babcock Forceps, Mid Grade, Sklar
Forceps
Surgi-OR™ Babcock Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue
Babcock Forceps are similar to Allis forceps; however, may be considered less traumatic due to their wider, rounded grasping surface. The jaws are circumferential, and the tips are triangular and fenestrated with horizontal serrations. They are particularly useful for grasping tube-shaped structures.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
They are used with intestinal and laparotomy procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty