Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sklarcut™ Ragnell Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
Sklarcut™
15-3320
89025-550EA 308.89 USD InStock
89025-550
Sklarcut™ Ragnell Dissecting Scissors, OR Grade, Sklar
Scissors
Sklarcut™ Ragnell Scissors, known as Kilner/Ragnell scissors are finger ring scissors, that feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue.
These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping, so the most accurate and clean cuts can be made. The long shanks and short jaws allow the user greater control with less effort during dissection. This combined with flat blades and blunt tips often make Ragnell scissors the dissection instrument of choice during longer procedures.
Sklarcut™ scissors are identified by a single gold coated finger ring on the handle of the serrated blade.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty