Would you like to log in again?
Sklarhone™ Plastic Surgery Scissors, OR Grade, Sklar®
Supplier: Sklar
15-3461 15-3520 15-3460
89025-598EA 209.19 USD InStock
89025-598 89025-600 89025-616
Sklarhone™ Plastic Surgery Scissors, OR Grade, Sklar®
Scissors
Sklarhone™ Plastic Surgery Scissors are non-ratcheted, finger ring scissors used for cutting and dissecting delicate tisssue in various plastic surgery procedures.
Sklarhone™ scissors feature one knife-like razor sharp edge and one standard scissor edge. This allows the user to make the sharp incisions of a knife with the cutting ability of scissors. Catching skin and pulls are minimized by the use of Sklarhone™ premium scissors. These instruments are identified by dual black finger rings.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty