Would you like to log in again?
Sklarcut™ Wagner Scissors, OR Grade, Sklar
Supplier: Sklar
Sklarcut™
15-3542
89025-626EA 282.13 USD InStock
89025-626
Sklarcut™ Wagner Scissors, OR Grade, Sklar
Scissors
Sklarcut™ Wagner Scissors are finger ring surgical scissors used in procedures requiring delicate or precise cutting, particularly in plastic surgery.
Sklarcut™ scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue. These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping, so the most accurate and clean cuts can be made.
Sklarcut™ Wagner Scissors are slightly longer than standard Wagner scissors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty