Would you like to log in again?
Sklarcut™ Iris Scissors, OR Grade, Sklar
Supplier: Sklar
Sklarcut™
15-3585 15-3590
89025-652EA 251.62 USD InStock
89025-652 89025-650
Sklarcut™ Iris Scissors, OR Grade, Sklar
Scissors
Sklarcut™ Iris Scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue
These fine, sharp scissors are designed for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures.
These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty