Would you like to log in again?
Sklar Edge™ Tungsten Carbide Wire Cutting Scissors, OR Grade, Sklar
Supplier: Sklar
Sklar Edge™
16-2800
89025-728EA 344.23 USD InStock
89025-728
Sklar Edge™ Tungsten Carbide Wire Cutting Scissors, OR Grade, Sklar
Scissors
Sklar Edge™ Tungsten Carbide Scissors provide a durable and hard cutting edge. They are designed to stay sharp for a longer period of time than standard scissors.
Sklar Edge™ TC Wire Cutting Scissors feature short shanks and very short jaws which are particularly useful for cutting wire sutures or wire mesh
Censitrac Ready products are industry standard and constructed of stainless steel.
The short jaws and small groove on each blade hold the wire and prevent it from slippling as pressure is applied for cutting.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty