Would you like to log in again?
LiteGrip™ Tungsten Carbide Adson Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
LiteGrip™
47-1448
89024-966EA 155.5 USD InStock
89024-966
LiteGrip™ Tungsten Carbide Adson Tissue Forceps, OR Grade, Sklar
Forceps
LiteGrip™ Tungsten Carbide Adson Tissue Forceps are for holding and manipulating delicate tissues.
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
LiteGrip™ instruments feature a lightweight, fenestrated handle. They are particularly useful during lengthy procedures when fatigue may arise. The finger holes on the handle also allow grip adjustments as needed by the surgeon.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty