Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glassy carbon, Powder 20-50 µm Type 1

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Warning
Synonyms: Vitreous carbon

041258-09 041258-18 041258-30
AA41258-09EA 56.99 USD InStock
AA41258-09 AA41258-18 AA41258-30
Glassy carbon, Powder 20-50 µm Type 1
Glassy carbon


Formula: C
MW: 12.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty