Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Graphite, plate, Thickness 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals

041831-GH
AA41831-GHEA 228.99 USD InStock
AA41831-GH
Graphite, plate, Thickness 6.35 mm (0.250 in)
Graphite
Ideal for casting and polishing purposes, moulding and carbon addition in steel industries, for smooth casting iron, copper base alloys and aluminum base alloy

  • Resin impregnated
  • Insoluble in water

It can be used for evaporating carbon onto specimens being prepared for either x-ray microanalysis (EDS or WDS) or transmission electron microscopy (TEM).


Formula: C
MW: 12.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty