Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium oxalate 0.1 N in aqueous solution analytical reagent

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium oxalateSodium ethanedioateSoerensen's buffer substances

047123-K2 47123-AK
AA47123-AKEA 72.37 USD InStock
AA47123-AK AA47123-K2
Disodium oxalate 0.1 N in aqueous solution analytical reagent
di-Sodium oxalate


Formula: C₂Na₂O₄
MW: 134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty