Would you like to log in again?
Stainless steel (Fe:Cr:Si:C; 82.9:16.5:0.5:0.02 wt%), powder -325 mesh Type 430-L
Supplier: Thermo Scientific Chemicals
Warning
47290-A3 047290-22 047290-36
AA47290-22EA 56.89 USD InStock
AA47290-22 AA47290-A3 AA47290-36
Stainless steel (Fe:Cr:Si:C; 82.9:16.5:0.5:0.02 wt%), powder -325 mesh Type 430-L
Stainless steel
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty