Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molecular sieve A4 (0.4 nm, 4 Å), pellets, Diameter 1-2mm (0.04-0.08in)

Supplier: Thermo Scientific Chemicals
Synonyms: Molecular sieve 0.4 nmMolecular sieves pack 4ASodium aluminium silicateLinde 4A

087956-A7 087956-A1 87956-A7 87956-30
AA87956-A1EA 148.39 USD InStock
CAAA87956-A7 AA87956-A1 AA87956-30 AA87956-A7
Molecular sieve A4 (0.4 nm, 4 Å), pellets, Diameter 1-2mm (0.04-0.08in)
Molecular sieve A4 (0.4 nm, 4 Å)


Storage Temperature: Ambient
MDL Number: MFCD00147627
CAS Number: 70955-01-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty