Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-cyanobutylzinc bromide 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger

Packaged under Argon in resealable ChemSeal™ bottles.

H58897-AD
AAH58897-ADEA 306.99 USD InStock
AAH58897-AD
4-cyanobutylzinc bromide 0.5 M in THF
4-cyanobutylzinc bromide


Formula: C₅H₈BrNZn
Density: 0.971
Storage Temperature: Ambient
MDL Number: MFCD01316996
CAS Number: 226570-68-9
UN: 2924
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty