Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Hydroxydiclofenac

Supplier: Thermo Scientific Chemicals
Danger

J64095-LB0
10814-314EA 627 USD InStock
10814-314
4'-Hydroxydiclofenac
4'-Hydroxydiclofenac
A Cytochrome P450 2C9 (CYP2C9) metabolite of diclofenac


Formula: C₁₄H₁₁Cl₂NO₃
Storage Temperature: Freezer
MDL Number: MFCD01671980
CAS Number: 64118-84-9
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty