Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Casein

Supplier: Thermo Scientific
J64214-36
10814-316EA 209.69 USD InStock
10814-316
Casein
Proteins and Peptides
CAS No.: 9000-71-9
Physical Form: Powder
Appearance: Off-white to pale yellow
Odor: Odorless
MDL No.: MFCD00081481

  • Hygroscopic

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty