Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cathepsin B Inhibitor 3

Supplier: Thermo Scientific
J65347-LB0
10814-382EA 382.42 USD InStock
10814-382
Cathepsin B Inhibitor 3
Proteins and Peptides
CAS No.: 134448-10-5
Molecular Formula: C18H29N3O6
Formula Weight: 383.44
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in methanol at 1mg/ml. Also soluble in DMSO or ethanol.
MDL No.: MFCD00797531

An inhibitor of Cathepsin B

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty