Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Formylphenyl-β-D-allopyranoside 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-(beta-D-Allopyranosyloxy)benzaldehydeHelicide

J66102-03 J66102-06
10814-440EA 66.24 USD InStock
10814-440 10814-438
4-Formylphenyl-β-D-allopyranoside 98%
4-Formylphenyl-beta-D-allopyranoside


Formula: C₁₃H₁₆O₇
MW: 284.26 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00210992
CAS Number: 80154-34-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty