Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gly-Arg-Gly-Asp-Ser-Pro

Supplier: Thermo Scientific
J66139-MCR
10814-450EA 48.59 USD InStock
10814-450
Gly-Arg-Gly-Asp-Ser-Pro
Proteins and Peptides
CAS No.: 91037-75-1
Molecular Formula: C22H37N9O10
Formula Weight: 587.58
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00076460

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty