Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[Nle4, D-Phe7] alpha-Melanocyte Stimulating Hormone amide Trifluoroacetate salt

Supplier: Thermo Scientific
J66191-MCR
10814-492EA 163.3 USD InStock
10814-492
[Nle4, D-Phe7] alpha-Melanocyte Stimulating Hormone amide Trifluoroacetate salt
Proteins and Peptides
CAS No.: 75921-69-6
Molecular Formula: C78H111N21O19
Formula Weight: 1647.00
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00151715

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty