Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Deoxy-D-glucosone ≥95%

Supplier: Thermo Scientific Chemicals

J66223-LB0
10814-522EA 220.11 USD InStock
10814-522
3-Deoxy-D-glucosone ≥95%
3-Deoxy-D-glucosone
Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents. Also used for cross-linking studies as it readily reacts with protein amino groups.


Formula: C₆H₁₀O₅
MW: 162.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01723101
CAS Number: 4084-27-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty