Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2,3,6-tri-O-benzoyl-ɑ-D-galactopyranoside 98%

Supplier: Thermo Scientific Chemicals

J66260-03 J66260-06
10814-548EA 192.76 USD InStock
10814-548 10814-546
Methyl-2,3,6-tri-O-benzoyl-ɑ-D-galactopyranoside 98%
Methyl-2,3,6-tri-O-benzoyl-alpha-D-galactopyranoside


Formula: C₂₈H₂₆O₉
MW: 506.51 g/mol
Melting Pt: 143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06200842
CAS Number: 3601-36-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty