Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ac-Leu-Glu-His-Asp-AMC

Supplier: Thermo Scientific Chemicals
Synonyms: N-Acetyl-Leu-Glu-His-Asp-7-amino-4-methylcoumarin

J66335-LB0 J66335-MCR
10815-566EA 147.1 USD InStock
10815-566 10815-568
Ac-Leu-Glu-His-Asp-AMC
Ac-Leu-Glu-His-Asp-AMC


Formula: C₃₃H₄₁N₇O₁₁
Storage Temperature: Freezer
MDL Number: MFCD03788104
CAS Number: 292633-16-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty