Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,5-Tri-O-benzyl-L-arabinofuranose 98%

Supplier: Thermo Scientific Chemicals

J66343-06 J66343-MD J66343-03
10814-560EA 42.78 USD InStock
10814-560 10814-558 10814-556
2,3,5-Tri-O-benzyl-L-arabinofuranose 98%
2,3,5-Tri-O-benzyl-L-arabinofuranose


Formula: C₂₆H₂₈O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00053542
CAS Number: 89615-42-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty