Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Growth Hormone Releasing Hexapeptide, Thermo Scientific

Supplier: Thermo Scientific
J66373-MB J66373-LB0
10814-592EA 302.31 USD InStock
10814-592 10814-590
Growth Hormone Releasing Hexapeptide, Thermo Scientific
Proteins and Peptides
CAS No.: 87616-84-0
Molecular Formula: C46H56N12O6
Formula Weight: 873.00
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00872694

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty