Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-Ser-Asp-Lys-Pro

Supplier: Thermo Scientific
J66430-LB0
10814-636EA 67.96 USD InStock
10814-636
N-Acetyl-Ser-Asp-Lys-Pro
Proteins and Peptides
CAS No.: 127103-11-1
Molecular Formula: C20H33N5O9
Formula Weight: 487.50
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Solubility: Soluble in water at 2mg/ml
MDL No.: MFCD00076849

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty