Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)

Supplier: Thermo Scientific
J66438-MCR
10814-642EA 167.39 USD InStock
10814-642
cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
Proteins and Peptides
CAS No.: 99534-03-9
Molecular Formula: C94H148N32O31
Formula Weight: 2222.40
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Solubility: Soluble in water
MDL No.: MFCD00133989

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty